Skip to Content
Merck
All Photos(2)

Key Documents

159638

Sigma-Aldrich

2,4-Dimethoxybenzyl alcohol

99%

Synonym(s):

2,4-Dimethoxybenzenemethanol, 2,4-Dimethoxyphenylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2OH
CAS Number:
Molecular Weight:
168.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

177-179 °C/10 mmHg (lit.)

mp

38-40 °C (lit.)

SMILES string

COc1ccc(CO)c(OC)c1

InChI

1S/C9H12O3/c1-11-8-4-3-7(6-10)9(5-8)12-2/h3-5,10H,6H2,1-2H3

InChI key

RNKOUSCCPHSCFE-UHFFFAOYSA-N

General description

2,4-Dimethoxybenzyl alcohol reacts with trifluoroacetic acid to yield calix[4]resorcinarene octamethyl ether, which on demethylation and acetylation yields the derived octa-acetate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

High yield synthesis of the parent C-unsubstituted calix [4] resorcinarene octamethyl ether.
Falana OM, et al.
Tetrahedron Letters, 35(1), 65-68 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service