Skip to Content
Merck
All Photos(1)

Key Documents

111287

Sigma-Aldrich

3-Penten-2-ol, predominantly trans

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH(OH)CH3
CAS Number:
Molecular Weight:
86.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.428 (lit.)

bp

119-121 °C (lit.)

density

0.843 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C=C\C(C)O

InChI

1S/C5H10O/c1-3-4-5(2)6/h3-6H,1-2H3/b4-3+

InChI key

GJYMQFMQRRNLCY-ONEGZZNKSA-N

General description

3-Penten-2-ol is an alcohol prepared by the inoculation of biofilms of M. mesophilicum with P. viridicatum.

Application

3-Penten-2-ol is a constituent of the kiwi fruit that can be analysed by multidimensional gas chromatography-olfactometry (GC-O).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L J Rose et al.
Current microbiology, 41(3), 206-209 (2000-07-29)
Volatile organic compounds from Penicillium viridicatum and Methylobacterium mesophilicum growing on laboratory media and on component materials of automobile air conditioners were analyzed with gas chromatography and mass spectrometry. P. viridicatum produced compounds such as 4-methyl thiazole, terpenes and alcohols
María J Jordán et al.
Journal of agricultural and food chemistry, 50(19), 5386-5390 (2002-09-05)
Gas chromatography-mass spectrometry (GC-MS) and multidimensional gas chromatography olfactometry (GC/GC-O) were utilized to study the aroma profile and the aroma active components of commercial kiwi essence and the initial fresh fruit puree. Totals of 29 and 33 components were identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service