Merck
  • New high-performance liquid chromatography method for the determination of (R)-warfarin and (S)-warfarin using chiral separation on a glycopeptide-based stationary phase.
New high-performance liquid chromatography method for the determination of (R)-warfarin and (S)-warfarin using chiral separation on a glycopeptide-based stationary phase.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2009-08-01)
Jana Malakova, Petr Pavek, Lucie Svecova, Iveta Jokesova, Pavel Zivny, Vladimir Palicka
ABSTRACT
Warfarin is a well-known anticoagulant agent that occurs in two enantiomers, (R)-(+)-warfarin and (S)-(-)-warfarin. A new liquid chromatography method for the determination of both enantiomers was developed, validated and applied in in vitro studies with the aim of evaluating the accumulation of (R)-warfarin and (S)-warfarin in the hepatoma HepG2 cell line. OptiMEM cell cultivation medium samples and cellular lysates were purified using Waters Oasis MAX extraction cartridges. The chiral separation of warfarin and the internal standard p-chlorowarfarin enantiomers was performed on an Astec Chirobiotic V2 column at a flow rate of 1.2mL/min. The mobile phase was composed of 31% acetonitrile, 5% of methanol and 64% of ammonium acetate buffer (10mmol/L, pH 4.1). The enantiomers were quantified using a fluorescence detector (lambda(excit)=320nm, lambda(emiss)=415nm). The limit of detection was found to be 0.121micromol/L of (S)-warfarin and 0.109micromol/L of (R)-warfarin. The range of applicability and linearity was estimated from 0.25 to 100micromol/L. The precision ranged from 1.3% to 12.2% of the relative standard deviation, and the accuracy reached acceptable values from 95.5% to 108.4%. The new bioanalytical method confirmed the same accumulation of (R)-warfarin and (S)-warfarin in the hepatoma HepG2 cell line.
Product Description
Supelco
Astec® CHIROBIOTIC® V2 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® V2 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V2 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V2 Chiral HPLC Guard Column, Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Astec® CHIROBIOTIC® V2 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Astec® CHIROBIOTIC® V2 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V2 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® V2 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm