Merck
Omics-Based Platform for Studying Chemical Toxicity Using Stem Cells.
Journal of proteome research (2017-12-21)
Yan Han, Jinghua Zhao, Ruili Huang, Menghang Xia, Daojing Wang
ABSTRACT
The new strategy for chemical toxicity testing and modeling is to use in vitro human cell-based assays in conjunction with quantitative high-throughput screening (qHTS) technology, to identify molecular mechanisms and predict in vivo responses. Stem cells are more physiologically relevant than immortalized cell lines because of their unique proliferation and differentiation potentials. We established a robust two stem cells-two lineages assay system, encompassing human mesenchymal stem cells (hMSCs) along osteogenesis and human induced pluripotent stem cells (hiPSCs) along hepatogenesis. We performed qHTS phenotypic screening of LOPAC1280 and identified 38 preliminary hits for hMSCs. This was followed by validation of a selected number of hits and determination of their IC
Sigma-Aldrich
NSC 95397, ≥97% (HPLC)
Sigma-Aldrich
Cell Counting Kit - 8, for quantitation of viable cell number in proliferation and cytotoxicity assays
Sigma-Aldrich
AIDA, solid
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Albumin antibody produced in mouse, clone HSA-11, ascites fluid
Sigma-Aldrich
Cantharidin
Sigma-Aldrich
SIGMAFAST BCIP®/NBT, tablet