Merck
  • The IKK-related kinase TBK1 activates mTORC1 directly in response to growth factors and innate immune agonists.
The IKK-related kinase TBK1 activates mTORC1 directly in response to growth factors and innate immune agonists.
The EMBO journal (2017-11-19)
Cagri Bodur, Dubek Kazyken, Kezhen Huang, Bilgen Ekim Ustunel, Kate A Siroky, Aaron Seth Tooley, Ian E Gonzalez, Daniel H Foley, Hugo A Acosta-Jaquez, Tammy M Barnes, Gabrielle K Steinl, Kae-Won Cho, Carey N Lumeng, Steven M Riddle, Martin G Myers, Diane C Fingar
ABSTRACT
The innate immune kinase TBK1 initiates inflammatory responses to combat infectious pathogens by driving production of type I interferons. TBK1 also controls metabolic processes and promotes oncogene-induced cell proliferation and survival. Here, we demonstrate that TBK1 activates mTOR complex 1 (mTORC1) directly. In cultured cells, TBK1 associates with and activates mTORC1 through site-specific mTOR phosphorylation (on S2159) in response to certain growth factor receptors (i.e., EGF-receptor but not insulin receptor) and pathogen recognition receptors (PRRs) (i.e., TLR3; TLR4), revealing a stimulus-selective role for TBK1 in mTORC1 regulation. By studying cultured macrophages and those isolated from genome edited mTOR S2159A knock-in mice, we show that mTOR S2159 phosphorylation promotes mTORC1 signaling, IRF3 nuclear translocation, and IFN-β production. These data demonstrate a direct mechanistic link between TBK1 and mTORC1 function as well as physiologic significance of the TBK1-mTORC1 axis in control of innate immune function. These data unveil TBK1 as a direct mTORC1 activator and suggest unanticipated roles for mTORC1 downstream of TBK1 in control of innate immunity, tumorigenesis, and disorders linked to chronic inflammation.
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Raptor Antibody, Upstate®, from rabbit
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Target shRNA Control Transduction Particles, Targets no known genes from any species
Sigma-Aldrich
Anti-phospho-mTOR (Ser2159) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
Polyinosinic–polycytidylic acid sodium salt, TLR ligand tested
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder