Merck
  • Effect of increased and maintained frequency of speed endurance training on performance and muscle adaptations in runners.
Effect of increased and maintained frequency of speed endurance training on performance and muscle adaptations in runners.
Journal of applied physiology (Bethesda, Md. : 1985) (2016-11-20)
Casper Skovgaard, Nicki Winfield Almquist, Jens Bangsbo
ABSTRACT
The aim of the study was, in runners accustomed to speed endurance training (SET), to examine the effect of increased and maintained frequency of SET on performance and muscular adaptations. After familiarization (FAM) to SET, 18 male (n = 14) and female (n = 4) runners (V̇o2max: 57.3 ± 3.4 ml/min; means ± SD) completed 20 sessions of maintained low-frequency (LF; every fourth day; n = 7) or high-frequency (HF; every second day; n = 11) SET. Before FAM as well as before and after an intervention period (INT), subjects completed a series of running tests and a biopsy from m. vastus lateralis was collected. Ten-kilometer performance improved (P < 0.05) ~3.5% during FAM with no further change during INT. Time to exhaustion at 90% vV̇o2max was 15 and 22% longer (P < 0.05) during FAM and a further 12 and 16% longer (P < 0.05) during INT in HF and LF, respectively. During FAM, muscle expression of NHE1 and maximal activity of citrate synthase (CS) and phosphofructokinase (PFK) increased (P < 0.05), running economy (RE) improved (P < 0.05), and V̇o2max was unchanged. During INT, both HF and LF increased (P < 0.05) muscle expression of NKAβ1, whereas maximal activity of CS and PFK, RE, and V̇o2max were unchanged. Furthermore, during INT, muscle expression of FXYD1 and SERCA1, and FXYD1 activity increased (P < 0.05) in HF, while muscle expression of SERCA2 decreased (P < 0.05) in LF. Thus increased or maintained frequency of SET leads to further improvements in short-term exercise capacity, but not in 10-km running performance. The better short-term exercise capacity may be associated with elevated expression of muscle proteins related to Na+/K+ transportation and Ca2+ reuptake. Ten speed endurance training (SET) sessions improved short-term exercise capacity and 10-km performance, which was followed by further improved short-term exercise capacity, but unchanged 10-km performance after 20 SET sessions performed with either high frequency (4 per 8 days) or continued low frequency (2 per 8 days) in trained runners. The further gain in short-term exercise capacity was associated with changes in muscle expression of proteins of importance for the development of fatigue.
Sigma-Aldrich
Anti-Na+/H+ Exchanger-1 Antibody, CT, clone 4E9, clone 4E9, Chemicon®, from mouse
Sigma-Aldrich
Anti-Na+K+ ATPase α-2 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution