Merck
  • Cardiac inflammation in genetic dilated cardiomyopathy caused by MYBPC3 mutation.
Cardiac inflammation in genetic dilated cardiomyopathy caused by MYBPC3 mutation.
Journal of molecular and cellular cardiology (2016-12-14)
Thomas L Lynch, Mohamed Ameen Ismahil, Anil G Jegga, Michael J Zilliox, Christian Troidl, Sumanth D Prabhu, Sakthivel Sadayappan
ABSTRACT
Cardiomyopathies are a leading cause of heart failure and are often caused by mutations in sarcomeric genes, resulting in contractile dysfunction and cellular damage. This may stimulate the production of a robust proinflammatory response. To determine whether myocardial inflammation is associated with cardiac dysfunction in dilated cardiomyopathy (DCM) caused by MYBPC3 mutation, we used the well-characterized cMyBP-C(t/t) mouse model of DCM at 3months of age. Compared to wild type (WT) mice, DCM mice exhibited significantly decreased fractional shortening (36.4±2% vs. 15.5±1.0%, p<0.0001) and significantly increased spleen weight (5.3±0.3 vs. 7.2±0.4mg/mm, p=0.002). Intriguingly, flow cytometry analysis revealed a significant increase in total (CD45+CD11b+Ly6C-MHCII+F480+) macrophages (6.5±1.4% vs. 14.8±1.4%, p=0.002) and classically activated (CD45+CD11b+Ly6C-MHCII+F480+CD206-) proinflammatory (M1) macrophages (3.4±0.8% vs. 10.3±1.2%, p=0.0009) in DCM hearts as compared with WT hearts. These results were further confirmed by immunofluorescence analysis of heart tissue sections. Splenic red pulp (CD11b+Ly6C+MHCIIlowF480hi) macrophages were significantly elevated (1.3±0.1% vs. 2.4±0.1%, p=0.0001) in DCM compared to WT animals. Serum cytokine analysis in DCM animals exhibited a significant increase (0.65±0.2 vs. 2.175±0.5pg/mL, p=0.02) in interleukin (IL)-6 compared to WT animals. Furthermore, RNA-seq analysis revealed the upregulation of inflammatory pathways in the DCM hearts. Together, these data indicate a robust proinflammatory response in DCM hearts, likely in response to cellular damage triggered by MYBPC3 mutation and resultant contractile dysfunction.
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
Donkey Anti-Rat IgG Antibody, Cy3 conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000