Merck
  • BRaf signaling principles unveiled by large-scale human mutation analysis with a rapid lentivirus-based gene replacement method.
BRaf signaling principles unveiled by large-scale human mutation analysis with a rapid lentivirus-based gene replacement method.
Genes & development (2017-04-14)
Chae-Seok Lim, Xi Kang, Vincent Mirabella, Huaye Zhang, Qian Bu, Yoichi Araki, Elizabeth T Hoang, Shiqiang Wang, Ying Shen, Sukwoo Choi, Bong-Kiun Kaang, Qiang Chang, Zhiping P Pang, Richard L Huganir, J Julius Zhu
ABSTRACT
Rapid advances in genetics are linking mutations on genes to diseases at an exponential rate, yet characterizing the gene-mutation-cell-behavior relationships essential for precision medicine remains a daunting task. More than 350 mutations on small GTPase
Sigma-Aldrich
2-Chloroadenosine
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 150,000-300,000
Sigma-Aldrich
DMH1, ≥98% (HPLC)
Sigma-Aldrich
Cesium methanesulfonate, used in patch clamp techniques