Merck
  • Synthesis of multiply substituted 1,6-dihydropyridines through Cu(I)-catalyzed 6-endo cyclization.
Synthesis of multiply substituted 1,6-dihydropyridines through Cu(I)-catalyzed 6-endo cyclization.
Organic & biomolecular chemistry (2015-05-01)
Haruki Mizoguchi, Ryo Watanabe, Shintaro Minami, Hideaki Oikawa, Hiroki Oguri
ABSTRACT
Copper-catalyzed 6-endo cyclization of N-propargylic β-enaminocarbonyls was developed for the synthesis of oxidation-labile 1,6-dihydropyridines. This synthetic method allows flexible and regio-defined assembly of various substituents at the N1, C2, C3, C4, and C6 positions of 1,6-dihydropyridines under mild conditions.
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
p-Anisaldehyde, natural, FG
Sigma-Aldrich
p-Anisaldehyde, 98%