Merck
  • Locomotor-reducing effects and structural characteristics of inhaled zerumbone and tetrahydrozerumbone derivatives.
Locomotor-reducing effects and structural characteristics of inhaled zerumbone and tetrahydrozerumbone derivatives.
Biological & pharmaceutical bulletin (2014-09-02)
Kakuyou Ogawa, Takashi Miyoshi, Takashi Kitayama, Michiho Ito
ABSTRACT
Zerumbone 1 is an 11-membered cyclic sesquiterpene obtained from the rhizomes of Zingiber zerumbet SMITH (Zingiberaceae). In this study, we investigated the structure-activity relationship of 1, α-humulene (2), tetrahydrozerumbone stereoisomers (3-5), and tetrahydrozerumbone derivatives (6-9). The oxygen-containing functional groups and the configurations at C1 and C2 contributed to the spontaneous locomotor activity reduction of zerumbone 1 and derivatives 2-9.
Product Description
Supelco
Triethyl citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethyl citrate, ≥99%, FCC, FG
Sigma-Aldrich
Triethyl citrate, ≥98.0% (GC)