Merck
  • Directing valvular interstitial cell myofibroblast-like differentiation in a hybrid hydrogel platform.
Directing valvular interstitial cell myofibroblast-like differentiation in a hybrid hydrogel platform.
Advanced healthcare materials (2014-06-25)
Jesper Hjortnaes, Gulden Camci-Unal, Joshua D Hutcheson, Sung Mi Jung, Frederick J Schoen, Jolanda Kluin, Elena Aikawa, Ali Khademhosseini
ABSTRACT
Three dimensional (3D) hydrogel platforms are powerful tools, providing controllable, physiologically relevant microenvironments that could aid in understanding how various environmental factors direct valvular interstitial cell (VIC) phenotype. Continuous activation of VICs and their transformation from quiescent fibroblast to activated myofibroblast phenotype is considered to be an initiating event in the onset of valve disease. However, the relative contribution VIC phenotypes is poorly understood since most 2D culture systems lead to spontaneous VIC myofibroblastic activation. Here, a hydrogel platform composed of photocrosslinkable versions of native valvular extracellular matrix components-methacrylated hyaluronic acid (HAMA) and methacrylated gelatin (GelMA)-is proposed as a 3D culture system to study VIC phenotypic changes. These results show that VIC myofibroblast-like differentiation occurs spontaneously in mechanically soft GelMA hydrogels. Conversely, differentiation of VICs encapsulated in HAMA-GelMA hybrid hydrogels, does not occur spontaneously and requires exogenous delivery of TGFβ1, indicating that hybrid hydrogels can be used to study cytokine-dependent transition of VICs. This study demonstrates that a hybrid hydrogel platform can be used to maintain a quiescent VIC phenotype and study the effect of environmental cues on VIC activation, which will aid in understanding pathobiology of valvular disease.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, ≥98%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O