Merck
Lactulose and renal failure.
Scandinavian journal of gastroenterology. Supplement (1997-01-01)
B Vogt, F J Frey
ABSTRACT
The introduction of lactulose as a new therapeutic agent for treatment of hepatic encephalopathy was a major breakthrough in this field. It was hypothesized that lactulose might prevent postoperative renal impairment after biliary surgery in patients with obstructive jaundice. The presumable mechanism purported was the diminished endotoxinemia by lactulose. Unfortunately, such a reno-protective effect has not been shown conclusively until now in clinical studies. In chronic renal failure lactulose is known to promote fecal excretion of water, sodium, potassium, amonium, urea, creatinine and protons. Thus, lactulose could be useful for the treatment of chronic renal failure. However, compliance to the therapy represents a major problem.
Product Description
Lactulose for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lactulose, ≥98.0% (HPLC)
Sigma-Aldrich
Lactulose, ≥95% (HPLC)
Lactulose, European Pharmacopoeia (EP) Reference Standard
USP
Lactulose, United States Pharmacopeia (USP) Reference Standard