Merck
  • The benefits and risks of using a levonorgestrel-releasing intrauterine system for contraception.
The benefits and risks of using a levonorgestrel-releasing intrauterine system for contraception.
Contraception (2011-11-10)
Diana Mansour
ABSTRACT
The contraceptive profile of the levonorgestrel-releasing intrauterine system (LNG-IUS; Mirena®) is well established, with efficacy similar to that achieved with sterilization and rapid return to fertility after discontinuation of use. The LNG-IUS is typically associated with transient menstrual disturbance during the first few months of use, but this usually settles with continued use, with a concomitant decrease in menstrual blood loss. Overall, the safety profile of the LNG-IUS has been well established across a wide population of women, and the available data do not suggest that the LNG-IUS adversely affects bone health or increase the risk of adverse cardiovascular events or breast and uterine cancers. This article reviews the literature to provide updated information on the risks and benefits associated with the LNG-IUS, particularly focusing on its use in contraception.
Product Description
Levonorgestrel, European Pharmacopoeia (EP) Reference Standard
USP
Levonorgestrel, United States Pharmacopeia (USP) Reference Standard
Supelco
D(−)-Norgestrel, analytical standard
Levonorgestrel for system suitability 2, European Pharmacopoeia (EP) Reference Standard