Merck
Valsartan: more than a decade of experience.
Drugs (2009-11-17)
Henry R Black, Jacqueline Bailey, Dion Zappe, Rita Samuel
ABSTRACT
Valsartan is a nonpeptide angiotensin receptor antagonist that selectively blocks the binding of angiotensin II to the angiotensin II type 1 receptor. The efficacy, tolerability and safety of valsartan have been demonstrated in large-scale studies in hypertension, heart failure (HF) and post-myocardial infarction (MI). This review focuses on what was learned from the valsartan clinical research programme and other comparative trials published from 1997 to the present. Many studies have demonstrated the efficacy of valsartan in lowering blood pressure (BP) in a variety of patient populations (including elderly, women, children, obese patients, patients with diabetes mellitus, patients with chronic kidney disease [CKD], patients at high risk of cardiovascular [CV] disease, African Americans, Hispanic Americans and Asians) and in improving outcomes in CV disease and CKD. In hypertension, valsartan exhibits dose-dependent efficacy in reducing both systolic and diastolic BP over the once-daily dose range of 80-320 mg; doses as high as 640 mg/day have been studied and found to be efficacious and safe. BP control can be enhanced with a more consistent 24-hour BP-lowering profile by using single-pill, fixed-dose combination therapy with valsartan plus hydrochlorothiazide (HCTZ). The cardioprotective benefits of valsartan have been demonstrated in large-scale outcome trials and include significant reductions in CV morbidity and mortality in HF, following MI, and in patients with co-morbid hypertension and coronary artery disease and/or HF; reductions in HF hospitalizations; and reductions in the incidence of stroke. The magnitude of these effects is comparable with that demonstrated with angiotensin-converting enzyme (ACE) inhibitors; however, valsartan has a more favourable tolerability profile, with a significantly lower incidence of cough and only rare reports of angio-oedema, both class effects of ACE inhibitor use. Consistent with its angiotensin receptor-blocking effects, valsartan also reduces circulating levels of biochemical markers that are associated with angiotensin II-mediated endothelial dysfunction and CV risk (e.g. high-sensitivity C-reactive protein or oxidized low-density lipoprotein). Improvements in CKD with valsartan include statistically and clinically meaningful reductions in urinary albumin and protein excretion in patients with type 2 diabetes and in nondiabetic patients with CKD. In short-term studies, valsartan has improved or stabilized various indices of metabolic function in at-risk patients, including those with co-morbid hypertension, obesity and/or metabolic syndrome. Because of this, valsartan is being prospectively investigated for its ability to reduce the incidence of new-onset diabetes and provide cardioprotection in patients with impaired glucose tolerance. Valsartan and valsartan/HCTZ are well tolerated. In clinical trials, adverse events during valsartan treatment were similar to those occurring with placebo. The combination of valsartan/HCTZ was better tolerated than HCTZ alone. Valsartan is administered once daily for hypertension; doses are usually taken upon awakening. In patients with HF or MI, valsartan is administered twice daily.
Product Description
Supelco
Valsartan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Valsartan, ≥98% (HPLC)
Valsartan, European Pharmacopoeia (EP) Reference Standard
Valsartan for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Valsartan, United States Pharmacopeia (USP) Reference Standard
Valsartan for system suitability, European Pharmacopoeia (EP) Reference Standard