Merck
  • Failure of thiamphenicol in a penicillin-allergic patient with Listeria meningoencephalitis--delayed cure following penicillin desensitization.
Failure of thiamphenicol in a penicillin-allergic patient with Listeria meningoencephalitis--delayed cure following penicillin desensitization.
Intensive care medicine (1992-01-01)
D Malvy, P H Dessalles, A Bailly, P Laffont, Y Monseau, J B Bonhoure
ABSTRACT
A 27-year-old woman, without compromised immunodefenses, experienced a Listeria meningoencephalitis, with brainstem symptoms. The identified agent exhibited poor susceptibility to usual effective antibiotics, except for penicillins. Knowledge of past history of an allergic reaction to beta-lactam antibiotics lead to appropriate therapy after acute intravenous desensitization of the patient to amoxicillin. Treatment resulted in therapeutic administration rate over 24 h, and in rapid regression of clinical and biological disorders.
Product Description
Thiamphenicol, European Pharmacopoeia (EP) Reference Standard
Supelco
Thiamphenicol, analytical standard
Sigma-Aldrich
Thiamphenicol