Merck
  • Development of novel potent orally bioavailable oseltamivir derivatives active against resistant influenza A.
Development of novel potent orally bioavailable oseltamivir derivatives active against resistant influenza A.
Journal of medicinal chemistry (2014-01-16)
Dennis Schade, Joscha Kotthaus, Lukas Riebling, Jürke Kotthaus, Helge Müller-Fielitz, Walter Raasch, Oliver Koch, Nora Seidel, Michaela Schmidtke, Bernd Clement
ABSTRACT
With the emergence of oseltamivir-resistant influenza viruses and in view of a highly pathogenic flu pandemic, it is important to develop new anti-influenza agents. Here, the development of neuraminidase (NA) inhibitors that were designed to overcome resistance mechanisms along with unfavorable pharmacokinetic (PK) properties is described. Several 5-guanidino- and 5-amidino-based oseltamivir derivatives were synthesized and profiled for their anti-influenza activity and in vitro and in vivo PK properties. Amidine 6 and guanidine 7 were comparably effective against a panel of different A/H1N1 and A/H3N2 strains and also inhibited mutant A/H1N1 neuraminidase. Among different prodrug strategies pursued, a simple amidoxime ethyl ester (9) exhibited a superior PK profile with an oral bioavailability of 31% (rats), which is comparable to oseltamivir (36%). Thus, bioisosteric replacement of the 5-guanidine with an acetamidine-in the form of its N-hydroxy prodrug-successfully tackled the two key limitations of currently used NA inhibitors, as exemplified with oseltamivir.
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution