Merck
  • Risk of selected cancers due to occupational exposure to chlorinated solvents in a case-control study in Montreal.
Risk of selected cancers due to occupational exposure to chlorinated solvents in a case-control study in Montreal.
Journal of occupational and environmental medicine (2012-11-14)
Krista Yorita Christensen, David Vizcaya, Harriet Richardson, Jérôme Lavoué, Kristan Aronson, Jack Siemiatycki
ABSTRACT
To evaluate the association between exposure to chlorinated solvents and cancer. We conducted a case-control study of occupational exposures and cancer in Montreal, Quebec, Canada, including 3730 cancer cases and 533 population controls. Occupational exposures were derived using a combination of subject-reported job history and expert assessment. We examined the associations between two chemical families and six chlorinated solvents with 11 sites of cancer. The majority of the associations examined were null, although many were based on small numbers. We found two significantly elevated odds ratios (ORs), one between perchloroethylene and prostate cancer (OR = 4.3; 95% CI: 1.4 to 13) and another between trichloroethylene and melanoma (OR = 3.2; 95% CI: 1.0 to 9.9). There was little evidence of associations between chlorinated solvents and cancer. Limited power precludes strong inferences about absence of risk. We raise hypotheses about two possible associations: perchloroethylene with prostate cancer and trichloroethylene with melanoma.
Product Description
Supelco
Density Standard 1623 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Tetrachloroethylene, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrachloroethylene, anhydrous, ≥99%
Sigma-Aldrich
Tetrachloroethylene, suitable for HPLC, ≥99.9%
Supelco
Tetrachloroethylene, analytical standard
Sigma-Aldrich
Tetrachloroethylene, ≥99.5%