Merck
A low-barrier hydrogen bond in the catalytic triad of serine proteases.
Science (New York, N.Y.) (1994-06-24)
P A Frey, S A Whitt, J B Tobin
ABSTRACT
Spectroscopic properties of chymotrypsin and model compounds indicate that a low-barrier hydrogen bond participates in the mechanism of serine protease action. A low-barrier hydrogen bond between N delta 1 of His57 and the beta-carboxyl group of Asp102 in chymotrypsin can facilitate the formation of the tetrahedral adduct, and the nuclear magnetic resonance properties of this proton indicate that it is a low-barrier hydrogen bond. These conclusions are supported by the chemical shift of this proton, the deuterium isotope effect on the chemical shift, and the properties of hydrogen-bonded model compounds in organic solvents, including the hydrogen bond in cis-urocanic acid, in which the imidazole ring is internally hydrogen-bonded to the carboxyl group.
Sigma-Aldrich
Dimethylmalonic acid, 98%
Supelco
Dimethylmalonic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland