Merck
  • Rhamnolipid (RL) from Pseudomonas aeruginosa OBP1: a novel chemotaxis and antibacterial agent.
Rhamnolipid (RL) from Pseudomonas aeruginosa OBP1: a novel chemotaxis and antibacterial agent.
Colloids and surfaces. B, Biointerfaces (2012-12-25)
P Bharali, J P Saikia, A Ray, B K Konwar
ABSTRACT
In the present study, the interaction of rhamnolipid produced by Pseudomonas aeruginosa OBP1 with the cell surfaces of Staphylococcus aureus (MTCC 3160) and Klebsiella pneumoniae (MTCC 618) were studied. Rhamnolipid concentration below critical micelle concentration (CMC) did not exhibit significant antibacterial activity. However, on increasing rhamnolipid concentration beyond CMC a prominent antibacterial activity was observed. The results demonstrated different degree of rhamnolipid interaction with both the bacteria. This might be due to the changes in their cell wall composition. The antibacterial activity determined by minimum inhibitory concentration (MIC). The antibacterial activity is prominent within 30 min of incubation. The antibacterial property of rhamnolipid was effective in all tested pH levels (5-9). The rhamnolipid was effective in almost all tested pH levels and showed better chemoattractant property against both the tested bacteria in comparison to glucose. The increase in the membrane permeability was evidenced by increase in the release of protein, enhancement in cell surface hydrophobicity and raises in the retention of crystal violet dye. Further, leakage of 260 nm absorbing intracellular materials, fourier transform infrared spectroscopy (FTIR) and scanning electron microscopy (SEM) analysis confirmed the disruptive action of rhamnolipid. The above evidences support the idea that rhamnolipid significantly alters the cell membrane/envelop that leads to cell damage and enhances membrane permeability. Such activity of rhamnolipid could be used as an additive in the formulation of antibiotic and other antimicrobial agents for enhancing the effectiveness of chemotherapeutics.
Product Description
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Supelco
Streptomycin sesquisulfate hydrate, VETRANAL®, analytical standard