Merck
  • Rosmarinic acid ameliorates acute liver damage and fibrogenesis in carbon tetrachloride-intoxicated mice.
Rosmarinic acid ameliorates acute liver damage and fibrogenesis in carbon tetrachloride-intoxicated mice.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-11-03)
Robert Domitrović, Marko Skoda, Vanja Vasiljev Marchesi, Olga Cvijanović, Ester Pernjak Pugel, Maja Bival Stefan
ABSTRACT
The aim of this study was to investigate the therapeutic potential of rosmarinic acid (RA), a natural phenolic, in the treatment of acute liver toxicity. RA at 10, 25 and 50mg/kg was administered by gavage once daily for 2 consecutive days, 6h after CCl(4) intoxication. CCl(4) intoxication caused hepatic necrosis and increased serum ALT activity. In the livers, oxidative/nitrosative stress was evidenced by increased 3-nitrotyrosine (3-NT) and thiobarbituric acid reactive substances (TBARS) formation and a significant decrease in Cu/Zn superoxide dismutase (SOD) activity. CCl(4) administration triggered inflammatory response in mice livers by activating nuclear factor-kappaB (NF-κB), which coincided with the induction of tumor necrosis factor-alpha (TNF-α) and cyclooxygenase-2 (COX-2). RA improved histological and serum markers of liver damage and significantly ameliorated oxidative/nitrosative stress and inflammatory response in liver tissue. Additionally, RA prevented transforming growth factor-beta1 (TGF-β1) and alpha-smooth muscle actin (α-SMA) expression, suggesting suppression of profibrotic response. Furthermore, RA significantly inhibited the CCl(4)-induced apoptosis, which was evident from decreased cleavage of caspase-3. The hepatoprotective activity of RA coincided with enhanced NF-E2-related factor 2 (Nrf2) and heme oxygenase-1 (HO-1) expression. The results of this study indicates that RA possesses antioxidant, anti-inflammatory, antiapoptotic and antifibrotic activity against acute liver toxicity.
Sigma-Aldrich
Rosmarinic acid, 96%
Rosmarinic acid, primary reference standard
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.