Merck
  • Comparison of different imidazolium supported ionic liquid polymeric phases with strong anion-exchange character for the extraction of acidic pharmaceuticals from complex environmental samples.
Comparison of different imidazolium supported ionic liquid polymeric phases with strong anion-exchange character for the extraction of acidic pharmaceuticals from complex environmental samples.
Journal of separation science (2012-08-07)
Dominika Bratkowska, Núria Fontanals, Sylwia Ronka, Andrzej W Trochimczuk, Francesc Borrull, Rosa M Marcé
ABSTRACT
Two imidazolium supported ionic liquid phases (SILPs) containing different anions, trifluoromethanesulphonate [CF(3)SO(3)(-)], and tetrafluoroborate [BF(4)(-)], were synthesized and evaluated as solid-phase extraction sorbents for extracting acidic pharmaceuticals from aqueous samples under strong anion-exchange conditions, which include an effective cleanup of the sample. The best SILP material [MI(+)][CF(3)SO(3)(-) ] was selected and successfully applied to the determination of acidic pharmaceuticals in different types of water samples (river water and effluent wastewater). The results were then compared to the previously synthesized SILP material based on [MI(+)][CF(3)COO(-)] and the commercially available Oasis MAX sorbent.
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%