Merck
  • Immobilization of lipase B within micron-sized poly-N-isopropylacrylamide hydrogel particles by solvent exchange.
Immobilization of lipase B within micron-sized poly-N-isopropylacrylamide hydrogel particles by solvent exchange.
Physical chemistry chemical physics : PCCP (2012-06-12)
Kornelia Gawlitza, Changzhu Wu, Radostina Georgieva, Dayang Wang, Marion B Ansorge-Schumacher, Regine von Klitzing
ABSTRACT
The aim of the present work is the use of a water soluble enzyme in an organic solvent, still with a pronounced catalytic activity. Therefore, lipase B from Candida antarctica (CalB) is immobilized within micron-sized thermosensitive p-NIPAM hydrogel particles using a solvent exchange from polar to organic solvents. The absorbed amount of CalB is investigated at different immobilization temperatures. Confocal laser scanning microscopy (CLSM) shows that CalB is homogeneously distributed within the polymer network. An enhanced specific activity of CalB in n-hexane is achieved after immobilization within the p-NIPAM microgels. In order to get information on the supply of the substrate depending on the temperature, the activity is determined at different reaction temperatures. Additionally, the system is stable in the organic solvent, namely n-hexane, and shows a good reusability.
Sigma-Aldrich
Poly(N-isopropyl acrylamide), NHS ester end functionalized, average Mn 5,000
Sigma-Aldrich
Poly(N-isopropylacrylamide), Mn ~40,000
Sigma-Aldrich
Poly(N-isopropylacrylamide), carboxylic acid terminated, average Mn 10,000