Merck
  • Structure-activity relationship study of N⁶-(2-(4-(1H-Indol-5-yl)piperazin-1-yl)ethyl)-N⁶-propyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine analogues: development of highly selective D3 dopamine receptor agonists along with a highly potent D2/D3 agonist and their pharmacological characterization.
Structure-activity relationship study of N⁶-(2-(4-(1H-Indol-5-yl)piperazin-1-yl)ethyl)-N⁶-propyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine analogues: development of highly selective D3 dopamine receptor agonists along with a highly potent D2/D3 agonist and their pharmacological characterization.
Journal of medicinal chemistry (2012-05-31)
Mark Johnson, Tamara Antonio, Maarten E A Reith, Aloke K Dutta
ABSTRACT
In our effort to develop multifunctional drugs against Parkinson's disease, a structure-activity-relationship study was carried out based on our hybrid molecular template targeting D2/D3 receptors. Competitive binding with [(3)H]spiroperidol was used to evaluate affinity (K(i)) of test compounds. Functional activity of selected compounds in stimulating [(35)S]GTPγS binding was assessed in CHO cells expressing either human D2 or D3 receptors. Our results demonstrated development of highly selective compounds for D3 receptor (for (-)-40K(i), D3 = 1.84 nM, D2/D3 = 583.2; for (-)-45K(i), D3 = 1.09 nM, D2/D3 = 827.5). Functional data identified (-)-40 (EC(50), D2 = 114 nM, D3 = 0.26 nM, D2/D3 = 438) as one of the highest D3 selective agonists known to date. In addition, high affinity, nonselective D3 agonist (-)-19 (EC(50), D2 = 2.96 nM and D3 = 1.26 nM) was also developed. Lead compounds with antioxidant activity were evaluated using an in vivo PD animal model.
Sigma-Aldrich
(2-Bromoethoxy)-tert-butyldimethylsilane, 99%
Sigma-Aldrich
Reserpine
Sigma-Aldrich
Ropinirole hydrochloride, powder, ≥98% (HPLC)
Supelco
Reserpine Standard for LC-MS, analytical standard, for LC-MS
Sigma-Aldrich
Reserpine, crystallized, ≥99.0% (HPLC)