Merck
  • Folding-promoted TBAX-mediated selective demethylation of methoxybenzene-based macrocyclic aromatic pentamers.
Folding-promoted TBAX-mediated selective demethylation of methoxybenzene-based macrocyclic aromatic pentamers.
Organic & biomolecular chemistry (2012-03-30)
Zhiyun Du, Bo Qin, Chang Sun, Ying Liu, Xi Zheng, Kun Zhang, Allan H Conney, Huaqiang Zeng
ABSTRACT
Described in this study is the ability of tetrabutylammonium salts (TBAX) to mediate an efficient mono- or di-demethylation removing one or two out of five aromatic methoxy methyl groups situated in similar chemical microenvironments in a H-bonded macrocyclic aromatic pentamer. These demethylations are found to be both chemo- and regioselective, and promoted by the H-bonding directed folding of the macrocyclic backbone.
Product Description
Supelco
Anisole, analytical standard
Sigma-Aldrich
Anisole, anhydrous, 99.7%
Sigma-Aldrich
Anisole, ≥99%, FCC, FG
Supelco
Anisole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anisole, ReagentPlus®, 99%