Merck
  • Spectrum of excess partial molar absorptivity. Part II: a near infrared spectroscopic study of aqueous Na-halides.
Spectrum of excess partial molar absorptivity. Part II: a near infrared spectroscopic study of aqueous Na-halides.
Physical chemistry chemical physics : PCCP (2012-02-24)
Fumie Sebe, Keiko Nishikawa, Yoshikata Koga
ABSTRACT
Our earlier thermodynamic studies suggested that F(-) and Cl(-) form hydration shells with the hydration number 14 ± 2 and 2.3 ± 0.6, respectively, and leave the bulk H(2)O away from hydration shells unperturbed. Br(-) and I(-), on the other hand, form hydrogen bonds directly with the momentarily existing hydrogen bond network of H(2)O, and retard the degree of entropy-volume cross fluctuation inherent in liquid H(2)O. The effect of the latter is stronger for I(-) than Br(-). Here we seek additional information about this qualitative difference between Cl(-) and (Br(-) and I(-)) pair by near infrared (NIR) spectroscopy. We analyze the ν(2) + ν(3) band of H(2)O in the range 4600-5500 cm(-1) of aqueous solutions of NaCl, NaBr and NaI, by a new approach. From observed absorbance, we calculate excess molar absorptivity, ε(E), excess over the additive contributions of solute and solvent. ε(E) thus contains information about the effect of inter-molecular interactions in the ν(2) + ν(3) spectrum. The spectrum of ε(E) shows three bands; two negative ones at 5263 and 4873 cm(-1), and the positive band at 5123 cm(-1). We then define and calculate the excess partial molar absorptivity of each salt, ε(E)(salt). From the behaviour of ε(E)(salt) we suggest that the negative band at 5263 cm(-1) represents free H(2)O without much hydrogen bonding under the influence of local electric field of ions. Furthermore, from a sudden change in the x(salt) (mole fraction of salt) dependence of ε(E)(salt), we suggest that there is an ion-pairing in x(salt) > 0.032, 0.036, and 0.04 for NaCl, NaBr and NaI respectively. The positive band of ε(E) at 5123 cm(-1) is attributed to a modestly organized hydrogen bond network of H(2)O (or liquid-likeness), and the x(salt) dependence of ε indicated a qualitative difference in the effect of Cl(-) from those of Br(-) and I(-). Namely, the values of ε(E)(salt) stay constant for Cl(-) but those for Br(-) and I(-) decrease smoothly on increasing the salt mole fraction. The mole fraction dependence of ε(E)(salt) at the 4873 cm(-1) band, due to ice-likeness in H(2)O, shows a subtle difference between Cl(-) and (Br(-), I(-)) pair.
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium iodide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Supelco
Density Standard 1251 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Sodium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium bromide, ReagentPlus®, ≥99%