Merck
  • Enantioselective organocatalytic three-component Petasis reaction among salicylaldehydes, amines, and organoboronic acids.
Enantioselective organocatalytic three-component Petasis reaction among salicylaldehydes, amines, and organoboronic acids.
Organic letters (2012-02-02)
Wen-Yong Han, Zhi-Jun Wu, Xiao-Mei Zhang, Wei-Cheng Yuan
ABSTRACT
The catalytic enantioselective three-component Petasis reaction among salicylaldehydes, amines, and organoboronic acids with a newly designed thiourea-binol catalyst is presented. A broad range of alkylaminophenols can be obtained in good yield (up to 92%) and good to high enantioselectivity (up to 95% ee). A possible reaction pathway for this catalytic enantioselective Petasis reaction is tentatively proposed.
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%
Supelco
Salicylaldehyde, analytical standard
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)