Merck
  • Induction of expression and functional activity of P-glycoprotein efflux transporter by bioactive plant natural products.
Induction of expression and functional activity of P-glycoprotein efflux transporter by bioactive plant natural products.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-08-20)
Alaa H Abuznait, Hisham Qosa, Nicholas D O'Connell, Jessica Akbarian-Tefaghi, Paul W Sylvester, Khalid A El Sayed, Amal Kaddoumi
ABSTRACT
The effect of bioactive plant natural products on the expression and functional activity of P-glycoprotein (P-gp) is poorly understood. Interactions of bioactive plant-based food and dietary supplements with P-gp can cause significant alteration of pharmacokinetic properties of P-gp substrate drugs when used in combination. This can augment toxicity and/or interfere with the drug's therapeutic outcomes. This study investigated the effects of diverse commonly used plant natural products on the expression and activity of P-gp in human adenocarcinoma cells (LS-180). These natural products included the tobacco cembranoid (1S,2E,4R,6R,7E,11E)-2,7,11-cembratriene-4,6-diol (cembratriene), the palm oil-derived γ-tocotrienol, the extra-virgin olive oil-derived secoiridoid oleocanthal, and the triterpene acid asiatic acid derived from Melaleuca ericifolia and abundant in several other common plant dietary supplements. Treatment with 25μM of cembratriene, oleocanthal, γ-tocotrienol, or asiatic acid showed 2.3-3.0-fold increase in P-gp expression as demonstrated by Western blotting. These results were consistent with those obtained by quantitative analysis of fluorescent micrographs for P-gp. Accumulation studies demonstrated 31-38% decrease in rhodamine 123 intracellular levels when LS-180 cells were treated with the investigated compounds as a result of P-gp induction. Bioactive natural products can up-regulate the P-gp expression and functionality, which may induce herb/food-drug interactions when concomitantly used with P-gp substrate drugs.
Sigma-Aldrich
Asiatic acid, ≥98% (HPLC), from Centella asiatica
Sigma-Aldrich
Asiatic acid, 97%