Merck
  • Improving the decolorization for textile dyes of a metagenome-derived alkaline laccase by directed evolution.
Improving the decolorization for textile dyes of a metagenome-derived alkaline laccase by directed evolution.
Applied microbiology and biotechnology (2011-04-28)
Yu Huan Liu, Mao Ye, Yi Lu, Xia Zhang, Gang Li
ABSTRACT
To obtain better performing laccases for textile dyes decolorization, random mutagenesis of Lac591, a metagenome-derived alkaline laccase, was carried out. After three rounds of error-prone PCR and high-throughput screening by assaying enzymatic activity toward the phenolic substrate 2,6-dimethoxyphenol (2,6-DMP), a mutant (Lac3T93) with remarkably improved enzymatic activity was obtained. Sequence analysis revealed that four amino acid substitutions (N40S, V55A, F62L, and E316V) were accumulated in the Lac3T93. Compared to the wild-type enzyme, the specific activity of Lac3T93 toward 2,6-DMP was increased to 4.8-fold (61.22 U/mg), and its optimal temperature and pH were changed to 60°C and 8.0 from 55°C and 7.5 of the wild-type enzyme, respectively. Furthermore, the degradation ability of Lac3T93 for textile dyes was investigated, and the new variant represented improved decolorization percentage for four industrial dyes with complex phenyl structure (Basic Blue 3, Methylene Blue, Bromophenol Blue, and Crystal Violet) and higher decolorization efficiency for Indigo Carmine than that of the parent enzyme. Furthermore, the decolorization percentage of Lac3T93 for five dyes in the absence of hydroxybenzotrizole (HBT) is clearly higher than those of the wild-type enzyme with 1 mM HBT, and HBT can further improve its decolorization ability.
Product Description
Supelco
2,6-Dimethoxyphenol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
2,6-Dimethoxyphenol, natural, ≥96%
Pricing and availability is not currently available.
Sigma-Aldrich
2,6-Dimethoxyphenol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
2,6-Dimethoxyphenol, ≥98%, FG
Pricing and availability is not currently available.