Merck
  • Nickel removal from nickel-5,10,15,20-tetraphenylporphine using supercritical water in absence of catalyst: a basic study.
Nickel removal from nickel-5,10,15,20-tetraphenylporphine using supercritical water in absence of catalyst: a basic study.
Journal of hazardous materials (2011-02-09)
Pradip Chandra Mandal, Wahyudiono, Mitsuru Sasaki, Motonobu Goto
ABSTRACT
Reactions of nickel-5,10,15,20-tetraphenylporphine (Ni-TPP) were studied in supercritical water in the presence of toluene without the addition of any catalyst, H(2) or H(2)S that is called a green process. The objective of this study was to remove nickel from Ni-TPP, the most common metal compound present in heavy crude, in high extent at low reaction time. All experiments were carried out in an 8.8 mL batch reactor fabricated from hastelloy C-276. The ability of supercritical water (SCW) to remove nickel from Ni-TPP was studied at temperatures of 450-490 °C and water partial pressures of 25-35 MPa. Water partial pressure had no effect on overall conversion at temperatures of 450 °C and a reaction time of 60 min. The overall Ni-TPP conversion was 89.80%, a figure above that of previous catalytic studies. The percentage of nickel removal was estimated as a function of reaction time and temperature. It were temperature 490 °C and pressure 25 MPa at reaction time 90 min where 65.68% nickel were removed by the action of SCW and toluene, as a co-solvent. It was determined that Ni-TPP undergoes a series of reactions, ending in demetallation and ring fragmentation. The obtained results suggest that supercritical water has a capability to remove nickel from Ni-TPP.
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, ≥99%
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, 97%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)