Merck
  • TBAF-catalysed silver oxide-mediated cross-coupling of functional trimethysilylpyridines: access to arylpyridines and bihetaryl compounds.
TBAF-catalysed silver oxide-mediated cross-coupling of functional trimethysilylpyridines: access to arylpyridines and bihetaryl compounds.
Organic & biomolecular chemistry (2011-01-26)
Frédéric Louërat, Heather Tye, Spencer Napier, Michael Garrigou, Mark Whittaker, Philippe C Gros
ABSTRACT
The concomitant use of silver oxide and catalytic amount of TBAF allowed the efficient and chemoselective coupling of readily available 4-chloro- and 4-methyl-2-trimethyl-silyl-pyridines with heteroaromatic and aromatic halides. Based on control experiments, a mechanism involving the formation of a pyridylsilver intermediate and TBAF recycling is postulated.
Sigma-Aldrich
Silver(I) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Silver oxide, predominantly silver(II) oxide
Sigma-Aldrich
Silver(I) oxide, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Silver(I) oxide, ReagentPlus®, 99%