Merck
  • Convergent synthesis of fluorescence-labeled probes of Annonaceous acetogenins and visualization of their cell distribution.
Convergent synthesis of fluorescence-labeled probes of Annonaceous acetogenins and visualization of their cell distribution.
Bioorganic & medicinal chemistry (2010-11-16)
Naoto Kojima, Takekuni Morioka, Daisuke Urabe, Masahiro Yano, Yuki Suga, Naoyoshi Maezaki, Ayako Ohashi-Kobayashi, Yasuyuki Fujimoto, Masatomo Maeda, Takao Yamori, Takehiko Yoshimitsu, Tetsuaki Tanaka
ABSTRACT
The convergent synthesis of fluorescence-labeled solamin, an antitumor Annonaceous acetogenin, was accomplished by two asymmetric alkynylations of 2,5-diformyl tetrahydrofuran with an alkyne tagged with fluorescent groups and another alkyne with an α,β-unsaturated γ-lactone. Assay for the growth inhibitory activity against human cancer cell lines revealed that the probe with the fluorescent groups at the end of the hydrocarbon chain may have the same mode of action as natural acetogenins. The merged fluorescence of dansyl-labeled solamin and MitoTracker Red suggests that Annonaceous acetogenins localize in the mitochondria.
Sigma-Aldrich
Benzethonium chloride, ≥97% (titration), ≥98% (HPLC)
Sigma-Aldrich
Benzethonium chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Hyamine® 1622 solution, 4 mM in H2O