Merck
  • Binding of 2-amino-4-chloro-m-benzenedisulfonamide as a metabolite of hydrochlorothiazide to erythrocytes.
Binding of 2-amino-4-chloro-m-benzenedisulfonamide as a metabolite of hydrochlorothiazide to erythrocytes.
Chemical & pharmaceutical bulletin (1990-10-01)
M Yamazaki, S Itoh, J Saji, K Tanabe, H Nakahama, Y Orita
ABSTRACT
2-Amino-4-chloro-m-benzenedisulfonamide (ACBS) is a metabolite of hydrochlorothiazide. We reported that the ACBS concentration in erythrocytes was higher than in plasma in a patient. Therefore the binding of ACBS to rabbit erythrocyte was studied. The Scatchard plot showed the nonlinear plot and the horizontal asymptote. Curvature in this plot indicated the existence of 2 classes of binding. One class was at a specific site, probably at carbonic anhydrase. Chromatographic data seemed to support the possibility.
Sigma-Aldrich
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%
Pricing and availability is not currently available.
Supelco
Benzothiadiazine Related Compound A, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.