Merck
  • Synthesis and application of calix[4]arene based resin for the removal of azo dyes.
Synthesis and application of calix[4]arene based resin for the removal of azo dyes.
Journal of hazardous materials (2009-07-31)
Muhammad Afzal Kamboh, Imam Bakhsh Solangi, S T H Sherazi, Shahabuddin Memon
ABSTRACT
The present study describes a novel synthetic method for the immobilization of calix[4]arene (II) onto the surface of modified Amberlite XAD-4 resin (4), which does not require the derivatization of calixarene moiety. The novel calix[4]arene based resin (C4 resin) 5 was used as sorbent for the removal of azo dyes. Batch-wise sorption study was carried out and observed that the C4 resin (5) is more effective as compared to compound II as well as pure Amberlite XAD-4 resin (1) to remove the selected dyes [i.e. Reactive Black-5 (RB-5), Reactive Red-45 (RR-45) and Congo Red (CR)]. The effect of sorbent dosage and pH on % sorption was studied. During the extraction process, various kinds of interactions such as electrostatic repulsion, deprotonation of the hydroxyl groups of C4 resin, dissociation of reactive dyes into anions/cations and structural variations were monitored and found that they are highly pH dependent.
Product Description
Supelco
Amberlite XAD4, 20-60 mesh
Supelco
Amberlite XAD4, 20-60 mesh
Supelco
Amberlite XAD4
Supelco
Amberlite XAD4, 20-60 mesh