Merck
  • Enantiodiscrimination of flexible cyclic solutes using NMR spectroscopy in polypeptide chiral mesophases: investigation of cis-decalin and THF.
Enantiodiscrimination of flexible cyclic solutes using NMR spectroscopy in polypeptide chiral mesophases: investigation of cis-decalin and THF.
The journal of physical chemistry. B (2009-07-16)
Christie Aroulanda, Olivier Lafon, Philippe Lesot
ABSTRACT
The conformational dynamics and orientational behavior of two model cyclic molecules, cis-decalin (cis-dec) and tetrahydrofurane (THF), dissolved in weakly ordering, polypeptidic chiral liquid crystals (CLCs) are theoretically discussed and experimentally investigated using deuterium and carbon-13 NMR spectroscopies. The analysis of enantiomeric and enantiotopic discriminations in these compounds is shown to depend on the rate of conformational exchange regime, slow or fast. The slow exchange regime is illustrated through the case of cis-dec at low temperature (243 K). We show that the deuterium NMR spectra in this regime can be qualitatively and quantitatively interpreted by restricting the conformational pathway of cis-dec to two enantiomeric conformers of C(2)-symmetry. The orientational order parameters of these interconverting enantiomers are calculated by matching the (2)H quadrupolar splittings with calculated conformer structures. The fast exchange regime is investigated through the examples of cis-dec at high temperature (356 K) and THF at room temperature (300 K). The (2)H NMR spectra above the coalescence temperature are analyzed by introducing the concept of "average molecular structure". This fictitious structure allows easily identifying NMR equivalences of solutes dissolved in CLC. However, it cannot be applied to determine consistent orientational order parameters. This study emphasizes that enantiotopic discriminations observed for flexible molecules in the fast exchange regime can be quantitatively interpreted only by considering the orientational order of each conformer.
Sigma-Aldrich
trans-Decahydronaphthalene, 99%
Sigma-Aldrich
cis-Decahydronaphthalene, 99%
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, reagent grade, 98%
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, anhydrous, ≥99%