Merck
  • Expedient preparation of all isomers of 2-aminocyclobutanecarboxylic acid in enantiomerically pure form.
Expedient preparation of all isomers of 2-aminocyclobutanecarboxylic acid in enantiomerically pure form.
The Journal of organic chemistry (2009-03-17)
Carlos Fernandes, Elisabeth Pereira, Sophie Faure, David J Aitken
ABSTRACT
A short, convenient, gram scale protocol has been established to allow facile access to all four stereoisomers of 2-aminocyclobutanecarboxylic acid, each in enantiomerically pure form (ee >99%). Starting from the readily available cis racemate, the procedure combines efficient alpha-phenylethylamine derivative resolution and controlled cis-to-trans epimerization procedures, and proceeds with invariably high yields.
Sigma-Aldrich
(S)-(−)-α-Methylbenzylamine, 98%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, purum, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
α-Methylbenzylamine, 99%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, 98%
Supelco
(R)-(+)-α-Methylbenzylamine, for chiral derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, ChiPros®, produced by BASF, ≥99.0%