Merck
  • Identification and quantitation of auxins in plants by liquid chromatography/electrospray ionization ion trap mass spectrometry.
Identification and quantitation of auxins in plants by liquid chromatography/electrospray ionization ion trap mass spectrometry.
Rapid communications in mass spectrometry : RCM (2008-07-26)
Qiaomei Lu, Lan Zhang, Tianwen Chen, Minghua Lu, Tong Ping, Guonan Chen
ABSTRACT
Auxin is an important phylohormone, which regulates specific physiological responses such as division, elongation and differentiation of cells. A new method using liquid chromatography/electrospray ionization ion trap mass spectrometry (LC/ESI-ITMS) has been developed for identification and quantitation of four auxins. Under the optimum conditions, four auxins (indole-3-acetic acid, indole-3-propionic acid, indole-3-butyric acid and 1-naphthylacetic acid) were completely separated and quantitated within 7 min with a minimum detection limit of 8.0 ng mL(-1) with relative standard deviations lower than 5.0%. This method also has been applied to analysis of auxins in Chinese cabbage where, even with a complicated serious background perturbation due to the natural biological matrix, the mean recoveries ranged from 77.5% to 99.8%. Finally, we discuss the MS-relevant properties of the identified auxins in detail.
Sigma-Aldrich
3-Indolepropionic acid, ReagentPlus®, 99%
Sigma-Aldrich
Indole-3-propionic acid, ≥99.0% (T)
Sigma-Aldrich
Indole-3-propionic acid, ≥97.0% (T)