Merck
  • Convergent total syntheses of fluvibactin and vibriobactin using molybdenum(VI) oxide-catalyzed dehydrative cyclization as a key step.
Convergent total syntheses of fluvibactin and vibriobactin using molybdenum(VI) oxide-catalyzed dehydrative cyclization as a key step.
Chemical communications (Cambridge, England) (2008-07-26)
Akira Sakakura, Shuhei Umemura, Kazuaki Ishihara
ABSTRACT
Efficient total syntheses of fluvibactin and vibriobactin have been achieved via molybdenum(VI) oxide-catalyzed dehydrative cyclization, Sb(OEt)(3)-catalyzed ester-amide transformation, and WSCI and HOAt-promoted dehydrative amide formation.
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%, suitable for plant cell culture
Sigma-Aldrich
Sodium molybdate, anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%
Sigma-Aldrich
Sodium molybdate dihydrate, 99.99% trace metals basis
Sigma-Aldrich
Sodium molybdate dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium molybdate, ≥98%