Merck
  • Synthesis and photophysics of a porphyrin-fullerene dyad assembled through Watson-Crick hydrogen bonding.
Synthesis and photophysics of a porphyrin-fullerene dyad assembled through Watson-Crick hydrogen bonding.
Chemical communications (Cambridge, England) (2005-03-30)
Jonathan L Sessler, Janarthanan Jayawickramarajah, Andreas Gouloumis, Tomas Torres, Dirk M Guldi, Stephen Maldonado, Keith J Stevenson
ABSTRACT
A novel porphyrin-fullerene dyad assembled through Watson-Crick hydrogen bonds is described; this system undergoes photoinduced electron transfer upon irradiation with visible light to produce a charge separated state that is substantially longer lived than that of previous dyads of this type.
Sigma-Aldrich
N-Methylaniline, 98%
Sigma-Aldrich
N-Methylaniline, ≥99%
Supelco
N-Methylaniline, analytical standard
Sigma-Aldrich
N-Methylaniline, purum, ≥98.0% (GC)