Merck
  • Decarboxylase activity involved in methyl ketone production by Staphylococcus carnosus 833, a strain used in sausage fermentation.
Decarboxylase activity involved in methyl ketone production by Staphylococcus carnosus 833, a strain used in sausage fermentation.
FEMS microbiology letters (2002-06-05)
S Fadda, A Lebert, S Leroy-Sétrin, R Talon
ABSTRACT
Staphylococcus carnosus strain 833, inoculated into sausage, increased the levels of methyl ketones which contributed to the cured aroma. These ketones were predicted to arise from incomplete beta-oxidation followed by a decarboxylation. To check this hypothesis, we measured the beta-decarboxylase activity in resting cells of S. carnosus grown in complex or in synthetic medium, using as substrate a beta-ketoacid, which can be an intermediate of the beta-oxidation pathway. This activity was present throughout the growth period. The enzyme appeared to be constitutive because no induction was observed. High aeration, a pH of 5 and the presence of nitrate promoted the production of methyl ketones.
Sigma-Aldrich
2-Pentanone, reagent grade, ≥90%
Sigma-Aldrich
2-Pentanone, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Pentanone, ≥98%, FCC, FG
Supelco
2-Pentanone, analytical standard
Sigma-Aldrich
2-Pentanone, ultrapure grade, ≥99.5%