Merck
  • Cortical brain organoid slices (cBOS) for the study of human neural cells in minimal networks.
Cortical brain organoid slices (cBOS) for the study of human neural cells in minimal networks.
iScience (2024-03-25)
Laura Petersilie, Sonja Heiduschka, Joel S E Nelson, Louis A Neu, Stephanie Le, Ruchika Anand, Karl W Kafitz, Alessandro Prigione, Christine R Rose
ABSTRACT
Brain organoids derived from human pluripotent stem cells are a promising tool for studying human neurodevelopment and related disorders. Here, we generated long-term cultures of cortical brain organoid slices (cBOS) grown at the air-liquid interphase from regionalized cortical organoids. We show that cBOS host mature neurons and astrocytes organized in complex architecture. Whole-cell patch-clamp demonstrated subthreshold synaptic inputs and action potential firing of neurons. Spontaneous intracellular calcium signals turned into synchronous large-scale oscillations upon combined disinhibition of NMDA receptors and blocking of GABAA receptors. Brief metabolic inhibition to mimic transient energy restriction in the ischemic brain induced reversible intracellular calcium loading of cBOS. Moreover, metabolic inhibition induced a reversible decline in neuronal ATP as revealed by ATeam1.03YEMK. Overall, cBOS provide a powerful platform to assess morphological and functional aspects of human neural cells in intact minimal networks and to address the pathways that drive cellular damage during brain ischemia.
Sigma-Aldrich
Anti-PSD95 Antibody, clone K28/43, clone K28/43, from mouse
Sigma-Aldrich
Anti-β-Tubulin III (neuronal) antibody, Mouse monoclonal, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, Cy3 conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
1(S),9(R)-(−)-Bicuculline methiodide, ≥95.0% (HPCE)
Sigma-Aldrich
Anti-Guinea Pig IgG (H+L), highly cross-adsorbed, CF 488A antibody produced in donkey, ~2 mg/mL, affinity isolated antibody