Merck
  • Neuronal STING activation in amyotrophic lateral sclerosis and frontotemporal dementia.
Neuronal STING activation in amyotrophic lateral sclerosis and frontotemporal dementia.
Acta neuropathologica (2024-03-13)
Christine Marques, Aaron Held, Katherine Dorfman, Joon Sung, Catherine Song, Amey S Kavuturu, Corey Aguilar, Tommaso Russo, Derek H Oakley, Mark W Albers, Bradley T Hyman, Leonard Petrucelli, Clotilde Lagier-Tourenne, Brian J Wainger
ABSTRACT
The stimulator of interferon genes (STING) pathway has been implicated in neurodegenerative diseases, including Parkinson's disease and amyotrophic lateral sclerosis (ALS). While prior studies have focused on STING within immune cells, little is known about STING within neurons. Here, we document neuronal activation of the STING pathway in human postmortem cortical and spinal motor neurons from individuals affected by familial or sporadic ALS. This process takes place selectively in the most vulnerable cortical and spinal motor neurons but not in neurons that are less affected by the disease. Concordant STING activation in layer V cortical motor neurons occurs in a mouse model of C9orf72 repeat-associated ALS and frontotemporal dementia (FTD). To establish that STING activation occurs in a neuron-autonomous manner, we demonstrate the integrity of the STING signaling pathway, including both upstream activators and downstream innate immune response effectors, in dissociated mouse cortical neurons and neurons derived from control human induced pluripotent stem cells (iPSCs). Human iPSC-derived neurons harboring different familial ALS-causing mutations exhibit increased STING signaling with DNA damage as a main driver. The elevated downstream inflammatory markers present in ALS iPSC-derived neurons can be suppressed with a STING inhibitor. Our results reveal an immunophenotype that consists of innate immune signaling driven by the STING pathway and occurs specifically within vulnerable neurons in ALS/FTD.
Sigma-Aldrich
RU.521, ≥98% (HPLC)
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-HuC/HuD Antibody, clone 15A7.1, clone 15A7.1, from mouse