Merck
  • Curative islet and hematopoietic cell transplantation in diabetic mice without toxic bone marrow conditioning.
Curative islet and hematopoietic cell transplantation in diabetic mice without toxic bone marrow conditioning.
Cell reports (2022-11-10)
Charles A Chang, Preksha Bhagchandani, Jessica Poyser, Brenda J Velasco, Weichen Zhao, Hye-Sook Kwon, Everett Meyer, Judith A Shizuru, Seung K Kim
ABSTRACT
Mixed hematopoietic chimerism can promote immune tolerance of donor-matched transplanted tissues, like pancreatic islets. However, adoption of this strategy is limited by the toxicity of standard treatments that enable donor hematopoietic cell engraftment. Here, we address these concerns with a non-myeloablative conditioning regimen that enables hematopoietic chimerism and allograft tolerance across fully mismatched major histocompatibility complex (MHC) barriers. Treatment with an αCD117 antibody, targeting c-Kit, administered with T cell-depleting antibodies and low-dose radiation permits durable multi-lineage chimerism in immunocompetent mice following hematopoietic cell transplant. In diabetic mice, co-transplantation of donor-matched islets and hematopoietic cells durably corrects diabetes without chronic immunosuppression and no appreciable evidence of graft-versus-host disease (GVHD). Donor-derived thymic antigen-presenting cells and host-derived peripheral regulatory T cells are likely mediators of allotolerance. These findings provide the foundation for safer bone marrow conditioning and cell transplantation regimens to establish hematopoietic chimerism and islet allograft tolerance.
Sigma-Aldrich
Anti-Guinea Pig IgG (H+L), CF 594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Guinea Pig IgG (H+L), CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody