Merck
  • Generation of transgenic Xenopus using restriction enzyme-mediated integration.
Generation of transgenic Xenopus using restriction enzyme-mediated integration.
Methods in molecular biology (Clifton, N.J.) (2012-06-13)
Mohammad Haeri, Barry E Knox
ABSTRACT
Transgenesis, the process of incorporating an exogenous gene (transgene) into an organism's genome, is a widely used tool to develop models of human diseases and to study the function and/or regulation of genes. Generating transgenic Xenopus is rapid and involves simple in vitro manipulations, taking advantage of the large size of the amphibian egg and external embryonic development. Restriction enzyme-mediated integration (REMI) has a number of advantages for transgenesis compared to other methods used to produce transgenic Xenopus, including relative efficiency, higher transgene expression levels, fewer genetic chimera in founder transgenic animals, and near-complete germ-line transgene transmission. This chapter explains the REMI method for generating transgenic Xenopus laevis tadpoles, including improvements developed to enable studies in the mature retina.
Sigma-Aldrich
Spermidine trihydrochloride, ≥98% (TLC)
Sigma-Aldrich
Chymostatin, microbial
Sigma-Aldrich
Leupeptin, Hemisulfate, Microbial, Leupeptin hemisulfate, CAS 103476-89-7, is a reversible inhibitor of trypsin-like proteases and cysteine proteases
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)