Merck
  • Semaphorin 4C promotes motility and immunosuppressive activity of cancer cells via CRMP3 and PD-L1.
Semaphorin 4C promotes motility and immunosuppressive activity of cancer cells via CRMP3 and PD-L1.
American journal of cancer research (2022-03-10)
Yu-Hsuan Hung, Ming-Derg Lai, Wen-Chun Hung, Li-Tzong Chen
ABSTRACT
Semaphorins (SEMAs) are membrane-bound or soluble proteins that participate in organ development and cancer progression, however, the detailed role of SEMAs in carcinogenesis is not fully elucidated yet. Our in silico analysis showed among the differentially expressed SEMAs in colon cancer tissues, patients with higher SEMA4C expression tumors had worse survival. The migration and invasion of the HCT116 and CT26 colon cancer cells were significantly suppressed by SEMA4C neutralizing antibody treatment; while enhanced by ectopic expression of SEMA4C. Subsequently, RNA sequencing study revealed microtubule polymerization- and nucleation-related genes are highly enriched in SEMA4C overexpression HCT116 cells. Western blotting showed the negative correlation between the levels of SEMA4C expression and tubulin acetylation. Mechanistic study showed SEMA4C interacted with and stabilized collapsin response mediator protein 3 (CRMP3), a novel deacetylase, to increase α-tubulin deacetylation and cell motility, which could be effectively attenuated after HDAC inhibitors treatment. We also found that a tumor-suppressive miRNA let-7b can target SEMA4C and act synergistically with SEMA4C neutralizing antibody to suppress the motility of colon cancer cells. In addition, blockade of SEMA4C could attenuate the expression of program death ligand 1 (PD-L1). Collectively, our results highlight that SEMA4C may promote colon cancer progression through modulating CRMP3-mediated tubulin deacetylation and PD-L1-mediated immunosuppression.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Protein G Mag Sepharose Xtra, Cytiva 28-9670-66, pack of 2 × 1 mL
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse