Merck
  • Mouse and human share conserved transcriptional programs for interneuron development.
Mouse and human share conserved transcriptional programs for interneuron development.
Science (New York, N.Y.) (2021-12-10)
Yingchao Shi, Mengdi Wang, Da Mi, Tian Lu, Bosong Wang, Hao Dong, Suijuan Zhong, Youqiao Chen, Le Sun, Xin Zhou, Qiang Ma, Zeyuan Liu, Wei Wang, Junjing Zhang, Qian Wu, Oscar Marín, Xiaoqun Wang
ABSTRACT
Genetic variation confers susceptibility to neurodevelopmental disorders by affecting the development of specific cell types. Changes in cortical and striatal γ-aminobutyric acid–expressing (GABAergic) neurons are common in autism and schizophrenia. In this study, we used single-cell RNA sequencing to characterize the emergence of cell diversity in the human ganglionic eminences, the transitory structures of the human fetal brain where striatal and cortical GABAergic neurons are generated. We identified regional and temporal diversity among progenitor cells underlying the generation of a variety of projection neurons and interneurons. We found that these cells are specified within the human ganglionic eminences by transcriptional programs similar to those previously identified in rodents. Our findings reveal an evolutionarily conserved regulatory logic controlling the specification, migration, and differentiation of GABAergic neurons in the human telencephalon.
Sigma-Aldrich
Anti-Vimentin Antibody, clone VIM 3B4, clone VIM 3B4, Chemicon®, from mouse
Sigma-Aldrich
Anti-FAM107A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Parvalbumin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit