Merck
Epigenetic state determines inflammatory sensing in neuroblastoma.
Proceedings of the National Academy of Sciences of the United States of America (2022-02-06)
Adam J Wolpaw, Liron D Grossmann, Jessica L Dessau, May M Dong, Bailey J Aaron, Patricia A Brafford, Darya Volgina, Guillem Pascual-Pasto, Alba Rodriguez-Garcia, Yasin Uzun, Marie Arsenian-Henriksson, Daniel J Powell, Kristopher R Bosse, Andrew Kossenkov, Kai Tan, Michael D Hogarty, John M Maris, Chi V Dang
ABSTRACT
Immunotherapy has revolutionized cancer treatment, but many cancers are not impacted by currently available immunotherapeutic strategies. Here, we investigated inflammatory signaling pathways in neuroblastoma, a classically "cold" pediatric cancer. By testing the functional response of a panel of 20 diverse neuroblastoma cell lines to three different inflammatory stimuli, we found that all cell lines have intact interferon signaling, and all but one lack functional cytosolic DNA sensing via cGAS-STING. However, double-stranded RNA (dsRNA) sensing via Toll-like receptor 3 (TLR3) was heterogeneous, as was signaling through other dsRNA sensors and TLRs more broadly. Seven cell lines showed robust response to dsRNA, six of which are in the mesenchymal epigenetic state, while all unresponsive cell lines are in the adrenergic state. Genetically switching adrenergic cell lines toward the mesenchymal state fully restored responsiveness. In responsive cells, dsRNA sensing results in the secretion of proinflammatory cytokines, enrichment of inflammatory transcriptomic signatures, and increased tumor killing by T cells in vitro. Using single-cell RNA sequencing data, we show that human neuroblastoma cells with stronger mesenchymal signatures have a higher basal inflammatory state, demonstrating intratumoral heterogeneity in inflammatory signaling that has significant implications for immunotherapeutic strategies in this aggressive childhood cancer.
Product Description
Millipore
MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A 48 Plex Premixed Magnetic Bead Panel - Immunology Multiplex Assay
Sigma-Aldrich
Human Serum, from human male AB plasma, USA origin, sterile-filtered