Merck
  • Persistence, dietary and ecological risk assessment of indoxacarb residue in/on tomato and soil using GC-MS.
Persistence, dietary and ecological risk assessment of indoxacarb residue in/on tomato and soil using GC-MS.
Food chemistry (2020-05-31)
T Sakthiselvi, M Paramasivam, D Vasanthi, K Bhuvaneswari
ABSTRACT
An efficient single quadrupole gas chromatography with mass spectrometry method was developed and validated for the determination of indoxacarb residues in tomato and soil. Residues were extracted from the samples using acetonitrile as extracting solvent and the extracts were purified through primary secondary amine and graphitized carbon black. Recoveries were obtained in the range of 92.12-110.51% with the relative standard deviation of 1.32-4.32%. Indoxacarb dissipated with half-life of 3.12-3.21 and 1.24-1.35d for tomato and soil, respectively following doses of indoxacarb 14.5% SC at 60, 90 and 120 g.a.i./ha. Safe waiting periods were found to be 1-3d. The residues were removed from tomato fruit was in the range of 16.73 to 54.32% using simple decontamination approaches. The present study suggest that the use of indoxacarb in tomato at recommended dose, does not seem to pose any dietary risk to the consumers. The soil RQ values indicated low level of risk to earthworms and arthropods.
Product Description
Supelco
Indoxacarb, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland