Merck
  • Alkyne- and Nitrile-Anchored Gold Nanoparticles for Multiplex SERS Imaging of Biomarkers in Cancer Cells and Tissues.
Alkyne- and Nitrile-Anchored Gold Nanoparticles for Multiplex SERS Imaging of Biomarkers in Cancer Cells and Tissues.
Nanotheranostics (2019-03-23)
Mingmin Li, Jianzhen Wu, Mengtian Ma, Zhichao Feng, Ze Mi, Pengfei Rong, Dingbin Liu
ABSTRACT
Surface-enhanced Raman spectroscopy (SERS) has proven a powerful tool for multiplex detection and imaging due to its narrow peak width and high sensitivity. However, conventional SERS reporters are limited to thiolated compounds, which have limitations such as chemical stability and spectral overlap. Here, we used alkyne- and nitrile-bearing molecules to directly fabricate a set of SERS tags for multiplex imaging. The alkyne and nitrile groups act as both the anchoring points to interact with gold nanoparticle (AuNP) surfaces and the reporters exhibiting strong and nonoverlapping signals in the cellular Raman-silent region. The SERS tags were subsequently modified with different antibodies for multicolor imaging of cancer cells and human breast cancer tissues. The reporters have a simple and readily accessible structure, hence providing new opportunities to prepare SERS nanoprobes.
Sigma-Aldrich
Gold nanoparticles, 5 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 80 nm diameter, alkyne functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 90 nm diameter, alkyne functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 10 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 100 nm diameter, Alkyne functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 30 nm diameter, Alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 70 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 15 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 20 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 60 nm diameter, Alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 40 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 50 nm diameter, Alkyne functionalized, 1 ML, OD 50, in water