Merck
A Novel Multisystem Proteinopathy Caused by a Missense ANXA11 Variant.
Annals of neurology (2021-05-29)
Tauana Bernardes Leoni, Carelis González-Salazar, Thiago Junqueira R Rezende, Ana Luisa C Hernández, Alexandre Hilário B Mattos, Antônio Rodrigues Coimbra Neto, Felipe Franco da Graça, João Pedro Nunes Gonçalves, Alberto R M Martinez, Lucas Taniguti, João Paulo Kitajima, Fernando Kok, Fábio Rogério, André Macedo Serafim da Silva, Alexandre Leite Rodrigues de Oliveira, Edmar Zanoteli, Anamarli Nucci, Marcondes C França
ABSTRACT
Protein misfolding plays a central role not only in amyotrophic lateral sclerosis (ALS), but also in other conditions, such as frontotemporal dementia (FTD), inclusion body myopathy (hIBM) or Paget's disease of bone. The concept of multisystem proteinopathies (MSP) was created to account for those rare families that segregate at least 2 out of these 4 conditions in the same pedigree. The calcium-dependent phospholipid-binding protein annexin A11 was recently associated to ALS in European pedigrees. Herein, we describe in detail 3 Brazilian families presenting hIBM (isolated or in combination with ALS/FTD) caused by the novel p.D40Y change in the gene encoding annexin A11 (ANXA11). We collected clinical, genetic, pathological and skeletal muscle imaging from 11 affected subjects. Neuroimaging was also obtained from 8 patients and 8 matched controls. Clinico-radiological phenotype of this novel hIBM reveals a slowly progressive predominant limb-girdle syndrome, but with frequent axial (ptosis/dropped head) and distal (medial gastrocnemius) involvement as well. Muscle pathology identified numerous rimmed vacuoles with positive annexin A11, TDP-43 and p62 inclusions, but no inflammation. Central nervous system was also involved: two patients had FTD, but diffusion tensor imaging uncovered multiple areas of cerebral white matter damage in the whole group (including the corticospinal tracts and frontal subcortical regions). These findings expand the phenotypic spectrum related to ANXA11. This gene should be considered the cause of a novel multisystem proteinopathy (MSP type 6), rather than just ALS. ANN NEUROL 2021;90:239-252.
Sigma-Aldrich
Monoclonal Anti-TARDBP antibody produced in mouse, clone 2E2-D3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-p62/SQSTM1 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution