Merck
  • Chronic T cell proliferation in brains after stroke could interfere with the efficacy of immunotherapies.
Chronic T cell proliferation in brains after stroke could interfere with the efficacy of immunotherapies.
The Journal of experimental medicine (2021-05-27)
Steffanie Heindl, Alessio Ricci, Olga Carofiglio, Qihui Zhou, Thomas Arzberger, Nikolett Lenart, Nicolai Franzmeier, Tibor Hortobagyi, Peter T Nelson, Ann M Stowe, Adam Denes, Dieter Edbauer, Arthur Liesz
ABSTRACT
Neuroinflammation is an emerging focus of translational stroke research. Preclinical studies have demonstrated a critical role for brain-invading lymphocytes in post-stroke pathophysiology. Reducing cerebral lymphocyte invasion by anti-CD49d antibodies consistently improves outcome in the acute phase after experimental stroke models. However, clinical trials testing this approach failed to show efficacy in stroke patients for the chronic outcome 3 mo after stroke. Here, we identify a potential mechanistic reason for this phenomenon by detecting chronic T cell accumulation-evading the systemic therapy-in the post-ischemic brain. We observed a persistent accumulation of T cells in mice and human autopsy samples for more than 1 mo after stroke. Cerebral T cell accumulation in the post-ischemic brain was driven by increased local T cell proliferation rather than by T cell invasion. This observation urges re-evaluation of current immunotherapeutic approaches, which target circulating lymphocytes for promoting recovery after stroke.
Sigma-Aldrich
Rose bengal, certified by the BSC, Dye content ≥80 %